Thermo Scientific Chemicals

Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals

Catalog number: 423300100
10 L, Each
Thermo Scientific Chemicals

Ammonium hydroxide, ACS reagent, 28-30% solution in water, Thermo Scientific Chemicals

Catalog number: 423300100
10 L, Each
Quantity
Catalog number: 423300100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1336-21-6
IUPAC Name
amine hydrate
Molecular Formula
H5NO
InChI Key
VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES
N.O
Appearance (Color)
Clear colorless
Appearance (Form)
Solution
Appearance
Free from suspended matter or sediment
Titration with HCl
28.0 to 30.0 % (as NH3)
Chloride (Cl)
=<0.5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text