Thermo Scientific Chemicals

Ammonium molybdate(VI) tetrahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423311000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium molybdate(VI) tetrahydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423311000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423311000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12054-85-2
IUPAC Name
ammonium hydrate molybdenum oxidandiide
Molecular Formula
H6MoNO2
InChI Key
XCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES
[NH4+].O.[O--].[Mo]
Appearance (Color)
White to off-white
Titration Complexometric
81.0 to 83.0 % (as MoO3)
Insoluble matter
=<0.005 %
Nitrate (NO3)
(Limit about 0.003%) Passes test
AsO4, PO4, SiO4 (as SiO2)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text