Phthalic anhydride, ACS reagent, Thermo Scientific Chemicals
Phthalic anhydride, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phthalic anhydride, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
500 g
2 kg
Packaging:
View all
Plastic Drum
Catalog number 423320050
Price (USD)/ Each
320.65
Online exclusive
356.00 
Save 35.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)/ Each
320.65
Online exclusive
356.00 
Save 35.35 (10%)
Add to cart
Phthalic anhydride, ACS reagent, Thermo Scientific Chemicals
Catalog number423320050
Price (USD)/ Each
320.65
Online exclusive
356.00 
Save 35.35 (10%)
-
Add to cart
Chemical Identifiers
CAS85-44-9
IUPAC Name1,3-dihydro-2-benzofuran-1,3-dione
Molecular FormulaC8H4O3
InChI KeyLGRFSURHDFAFJT-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.003 %
Appearance (Color)White
Appearance (Form)Flakes
Infrared spectrumConforms
Melting point129°C to 132°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only