Thermo Scientific Chemicals

Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals

Catalog number: 423330250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals

Catalog number: 423330250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 423330250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13573-16-5
IUPAC Name
chromium(3+) ammonium tetrakis(cyanosulfanide) diamine
Molecular Formula
C4H10CrN7S4
InChI Key
ZGLIQORZYPZFPW-UHFFFAOYSA-K
SMILES
N.N.[NH4+].[Cr+3].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
Appearance (Color)
Red
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Gravimetry
>=93.0 %
Insoluble matter
=<0.05 % (in dilute HCl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text