Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals
Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Reinecke Salt, ACS Reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 423331000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namechromium(3+) ammonium tetrakis(cyanosulfanide) diamine
Molecular FormulaC4H10CrN7S4
InChI KeyZGLIQORZYPZFPW-UHFFFAOYSA-K
SMILESN.N.[NH4+].[Cr+3].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N
Molecular Weight (g/mol)336.41
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Gravimetry>=93.0 %
Insoluble matter=<0.05 % (in dilute HCl)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only