Thermo Scientific Chemicals

Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content

Catalog number: 423340010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Acridine Orange, hydrochloride hydrate, pure, ca. 80 % dye content

Catalog number: 423340010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 423340010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1704465-79-1
IUPAC Name
N3,N3,N6,N6-tetramethylacridine-3,6-diamine hydrochloride
Molecular Formula
C17H20ClN3
InChI Key
VSTHNGLPHBTRMB-UHFFFAOYSA-N
SMILES
Cl.CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
UV spectrophotometry
>=75 %
Loss on drying
=<5 % (105°C)
Appearance (Color)
Orange to brown
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text