Ammonium nitrate, 98%, ACS reagent, Thermo Scientific Chemicals
Ammonium nitrate, 98%, ACS reagent, Thermo Scientific Chemicals
Ammonium nitrate, 98%, ACS reagent, Thermo Scientific Chemicals
Ammonium nitrate, 98%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium nitrate, 98%, ACS reagent, Thermo Scientific Chemicals

Quantity:
1 kg
25 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 423350010
Price (JPY)
-
Request A Quote
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS6484-52-2
IUPAC Nameammonium nitrate
Molecular FormulaH4N2O3
InChI KeyDVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES[NH4+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals
Titration excess NaOH>=97.5 %
Heavy metals (as Pb)=<5 ppm
Residue after ignition=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link