Ammonium oxalate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Ammonium oxalate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium oxalate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number 423360025
Price (USD)/ Each
-
Quantity:
2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS6009-70-7
IUPAC Nameoxalic acid diamine hydrate
Molecular FormulaC2H10N2O5
InChI KeyMSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILESN.N.O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with KMnO499.0 to 101.0 %
Heavy metals (as Pb)=<5 ppm
Insoluble matter=<0.005 %
Residue after ignition=<0.02 %
Chloride (Cl)=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only