Thermo Scientific Chemicals

Ammonium oxalate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423361000
100 g, Each
Thermo Scientific Chemicals

Ammonium oxalate monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423361000
100 g, Each
Quantity
Catalog number: 423361000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6009-70-7
IUPAC Name
oxalic acid diamine hydrate
Molecular Formula
C2H10N2O5
InChI Key
MSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILES
N.N.O.OC(=O)C(O)=O
Titration with KMnO4
99.0 to 101.0 %
Heavy metals (as Pb)
=<5 ppm
Insoluble matter
=<0.005 %
Residue after ignition
=<0.02 %
Chloride (Cl)
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text