Thermo Scientific Chemicals

Ammonium sulfamate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423390025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Ammonium sulfamate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423390025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 423390025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7773-06-0
IUPAC Name
ammonium sulfamate
Molecular Formula
H6N2O3S
InChI Key
GEHMBYLTCISYNY-UHFFFAOYSA-N
SMILES
[NH4+].NS([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Melting point
132.0°C to 134.0°C
Titration
>=98.0 %
Heavy metals (as Pb)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text