Aniline, 99+%, ACS reagent, Thermo Scientific Chemicals
Aniline, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aniline, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
500 mL
1 L
Packaging:
View all
Glass bottle
Catalog number 423420025
Price (USD)/ Each
347.00
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
347.00
Add to cart
Aniline, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number423420025
Price (USD)/ Each
347.00
-
Add to cart
Chemical Identifiers
IUPAC Nameaniline
Molecular FormulaC6H7N
InChI KeyPAYRUJLWNCNPSJ-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1
Molecular Weight (g/mol)93.13
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5845 to 1.5875 (20°C, 589 nm)
Chlorobenzene (C6H5Cl)=<0.01 %
Nitrobenzene (C6H5NO2)(Limit about 0.001 %) Passes test
Appearance (Form)Clear liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only