Thermo Scientific Chemicals

Barium acetate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423435000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Barium acetate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423435000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423435000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
543-80-6
IUPAC Name
barium(2+) diacetate
Molecular Formula
C4H6BaO4
InChI Key
ITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES
[Ba++].CC([O-])=O.CC([O-])=O
Appearance (Form)
Crystalline powder
Oxidizing substance (as NO3)
=<0.005 %
Appearance (Color)
White
Heavy metals (as Pb)
=<5 ppm
Titration Complexometric
99.0 to 102.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text