Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423450025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Barium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423450025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423450025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10326-27-9
IUPAC Name
barium(2+) dihydrate dichloride
Molecular Formula
BaCl2H4O2
InChI Key
PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ba++]
Appearance (Form)
Crystalline powder or crystals
Titration Complexometric
>=99.0 %
Heavy metals (ICP-OES)
=<5 ppm
pH
5.2 to 8.2 (5 % at 25.0°C)
Insoluble matter
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text