Benzoic acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Benzoic acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzoic acid, 99.6%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
25 g
500 g
Packaging:
Plastic bottle
Catalog number 423470020
Price (USD)/ Each
-
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS65-85-0
IUPAC Namebenzoic acid
Molecular FormulaC7H6O2
InChI KeyWPYMKLBDIGXBTP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Freezing point122°C to 123°C
Titration with NaOH>=99.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only