Thermo Scientific Chemicals

1-Butanol, 99.5%, for analysis ACS, Thermo Scientific Chemicals

Catalog number: 423490025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

1-Butanol, 99.5%, for analysis ACS, Thermo Scientific Chemicals

Catalog number: 423490025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 423490025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-36-3
IUPAC Name
butan-1-ol
Molecular Formula
C4H10O
InChI Key
LRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILES
CCCCO
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=99.4 %
Water
=<0.1 % (K.F.)
Density
(20/20°C) 0.809 to 0.812

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text