1-Butanol, 99.5%, for analysis ACS, Thermo Scientific Chemicals
1-Butanol, 99.5%, for analysis ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Butanol, 99.5%, for analysis ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
500 mL
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number 423490100
Price (USD)/ Each
-
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.4 %
Water=<0.1 % (K.F.)
Density(20/20°C) 0.809 to 0.812
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only