Thermo Scientific Chemicals

Calcium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423520250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423520250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423520250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10035-04-8
IUPAC Name
calcium dihydrate dichloride
Molecular Formula
CaCl2H4O2
InChI Key
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ca++]
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals
pH
4.5 to 8.5 (5 % at 25°C)
Insoluble matter
=<0.01 %
Oxidizing substance (as NO3)
=<0.003 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text