Calcium nitrate tetrahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Calcium nitrate tetrahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423530250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS13477-34-4
IUPAC Namecalcium bis(nitric acid) tetrahydrate
Molecular FormulaCaH10N2O10
InChI KeyDKCJOKYHYIAZQH-UHFFFAOYSA-N
SMILESO.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Complexometric99.0 to 103.0 %
Insoluble matter=<0.005 %
Barium (Ba)=<50 ppm
Magnesium (Mg)=<500 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only