Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Thermo Scientific Chemicals

Citric acid, ≥99.5%, ACS reagent, anhydrous

CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 423560020
Price (EUR)
79,22
Online Exclusive
93,20
Save 13,98 (15%)
Each
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
79,22
Online Exclusive
93,20
Save 13,98 (15%)
Each
Add to cart
Chemical Identifiers
CAS77-92-9
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular FormulaC6H8O7
InChI KeyKRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILESOC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=99.5 %
Carboniz. sub. by hot H2SO4(tartrates, etc) Passes test
Chloride (Cl)=<10 ppm
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

EU Specific Hazard Statements

May form combustible dust concentrations in air

Precautionary Statements

P280 - Wear eye protection/ face protection

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link