Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423571000
100 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423571000
100 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 423571000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-13-1
IUPAC Name
λ²-cobalt(2+) hexahydrate dichloride
Molecular Formula
Cl2CoH12O6
InChI Key
GFHNAMRJFCEERV-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Co++]
Appearance (Color)
Violet
Titration Complexometric
98.0 to 102.0 %
Iron (Fe)
=<50 ppm
Zinc (Zn)
=<300 ppm
Nitrate (NO3)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text