Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423595000
Price (USD)/ Each
102.00
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
102.00
Add to cart
Copper(II) acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
Catalog number423595000
Price (USD)/ Each
102.00
-
Add to cart
Chemical Identifiers
CAS6046-93-1
IUPAC Namecopper(2+) diacetate hydrate
Molecular FormulaC4H8CuO5
InChI KeyNWFNSTOSIVLCJA-UHFFFAOYSA-L
SMILESO.[Cu++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue-green to dark green
Appearance (Form)Solid
Infrared spectrumConforms
Titration Iodometric98.0 to 102.0 %
Chloride (Cl)=<30 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only