Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423635000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Dimethylaminobenzaldehyde, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423635000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 423635000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
100-10-7
IUPAC Name
4-(dimethylamino)benzaldehyde
Molecular Formula
C9H11NO
InChI Key
BGNGWHSBYQYVRX-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=O)C=C1
Appearance (Color)
Light yellow to light green
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Residue after ignition
=<0.1 %
Solubility
(in alcohol) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text