Diphenylamine, ACS reagent, Thermo Scientific Chemicals
Diphenylamine, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diphenylamine, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 423651000
Price (USD)/ Each
64.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
75.80 
Save 11.15 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
75.80 
Save 11.15 (15%)
Add to cart
Diphenylamine, ACS reagent, Thermo Scientific Chemicals
Catalog number423651000
Price (USD)/ Each
64.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
75.80 
Save 11.15 (15%)
-
Add to cart
Chemical Identifiers
CAS122-39-4
IUPAC NameN-phenylaniline
Molecular FormulaC12H11N
InChI KeyDMBHHRLKUKUOEG-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals or flakes
Infrared spectrumConforms
Melting point52.5°C to 54.0°C
Residue after ignition=<0.03 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only