Thermo Scientific Chemicals

Ethyl Acetate, 99.6%, For analysis ACS, Thermo Scientific Chemicals

Catalog number: 423680250
25 L, Metal Drum, Each
Thermo Scientific Chemicals

Ethyl Acetate, 99.6%, For analysis ACS, Thermo Scientific Chemicals

Catalog number: 423680250
25 L, Metal Drum, Each
Quantity
Packaging
Catalog number: 423680250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
141-78-6
IUPAC Name
ethyl acetate
Molecular Formula
C4H8O2
InChI Key
XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES
CCOC(C)=O
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=99.5 %
Water
=<0.2 % (K.F.)
Density
(20/20°C) 0.901 to 0.904

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text