Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423690250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423690250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423690250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-83-7
IUPAC Name
iron(3+) ammonium dodecahydrate disulfate
Molecular Formula
FeH28NO20S2
InChI Key
LCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Nitrate (NO3)
=<0.01 %
Appearance (Form)
Adhering powder or crystals
Appearance (Color)
Violet
Titration Iodometric
98.5 to 102.0 %
Ferrous iron (Fe 2+)
(Limit about 0.001%) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text