Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, 97+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423705000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Iron(III) chloride hexahydrate, 97+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423705000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 423705000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10025-77-1
IUPAC Name
iron(3+) hexahydrate trichloride
Molecular Formula
Cl3FeH12O6
InChI Key
NQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
Appearance (Color)
Yellow to orange to light brown
Appearance (Form)
Adhering powder and/or lumps
Titration Iodometric
97.0 to 102.0 %
Insoluble matter
=<0.01 %
Phosphorous compounds (as PO4)
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text