Thermo Scientific Chemicals

Iron(III) nitrate nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423715000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Iron(III) nitrate nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423715000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423715000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-61-8
IUPAC Name
iron(3+) nonahydrate trinitrate
Molecular Formula
FeH18N3O18
InChI Key
SZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Magnesium (Mg)
=<50 ppm
Chloride (Cl)
=<5 ppm
Sulfate (SO4)
=<100 ppm
Appearance (Color)
Almost white to light grey to light purple
Appearance (Form)
Adhering crystals or chunks

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text