Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
3 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 423731000
Price (USD)/ Each
33.65
Online exclusive
39.70 
Save 6.05 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
33.65
Online exclusive
39.70 
Save 6.05 (15%)
Add to cart
Iron(II) sulfate heptahydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number423731000
Price (USD)/ Each
33.65
Online exclusive
39.70 
Save 6.05 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameλ²-iron(2+) heptahydrate sulfate
Molecular FormulaFeH14O11S
InChI KeySURQXAFEQWPFPV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)278.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to blue-green to light blue
Appearance (Form)Crystals
Titration with KMnO4>=99.0 %
Insoluble matter=<0.01 %
Phosphate (PO4)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only