Thermo Scientific Chemicals

Hexanes, 99%, ACS reagent, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 423760025
2.5 L, Glass Bottle, Each
Thermo Scientific Chemicals

Hexanes, 99%, ACS reagent, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 423760025
2.5 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 423760025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92112-69-1
IUPAC Name
hexane
Molecular Formula
C6H14
InChI Key
VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES
CCCCCC
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=98.5 % Sum of 5 isomers + total Hexanes
GC
+ Methylcyclopentane
Residue after evaporation
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text