Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 423770500
Price (USD)/ Each
41.65
Online exclusive
48.70 
Save 7.05 (14%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
48.70 
Save 7.05 (14%)
Add to cart
Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals
Catalog number423770500
Price (USD)/ Each
41.65
Online exclusive
48.70 
Save 7.05 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namehydrazine; sulfuric acid
Molecular FormulaH6N2O4S
InChI KeyZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILESNN.OS(O)(=O)=O
Molecular Weight (g/mol)130.12
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<50 ppm
Iron (Fe)=<10 ppm
Appearance (Form)Crystalline powder
Titration Iodimetric>=99.0 %
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only