Thermo Scientific Chemicals

Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423771000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Hydrazine sulfate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423771000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423771000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-93-2
IUPAC Name
hydrazine; sulfuric acid
Molecular Formula
H6N2O4S
InChI Key
ZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILES
NN.OS(O)(=O)=O
Appearance (Color)
White
Chloride (Cl)
=<50 ppm
Iron (Fe)
=<10 ppm
Appearance (Form)
Crystalline powder
Titration Iodimetric
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text