Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals

Hydrobromic acid, 47.0 to 49.0%, BrH, CAS Number-10035-10-6, 7732-18-5, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, 126.0 deg.C to 128.0 deg.C, CHEBI:47266
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 423785000
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
Add to cart
Hydrobromic Acid, Reagent ACS, Thermo Scientific Chemicals
Catalog number423785000
Price (USD)/ Each
122.65
Online exclusive
136.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10035-10-6
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
Titration with NaOH47.0 to 49.0 %
Heavy metals (ICP-OES)=<5 ppm
Residue after ignition=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only