Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals
Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 423800250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7664-39-3
IUPAC Namehydrogen fluoride
Molecular FormulaFH
InChI KeyKRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILESF
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
Titration with NaOH48.0 to 51.0 %
Heavy metals (as Pb)=<0.5 ppm
Residue after ignition=<0.0005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only