Thermo Scientific Chemicals

Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals

Catalog number: 423805000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Hydrofluoric acid, ACS reagent, 48-51% solution in water, Thermo Scientific Chemicals

Catalog number: 423805000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423805000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7664-39-3
IUPAC Name
hydrogen fluoride
Molecular Formula
FH
InChI Key
KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES
F
Appearance (Color)
Clear colorless
Appearance (Form)
Solution
Titration with NaOH
48.0 to 51.0 %
Heavy metals (as Pb)
=<0.5 ppm
Residue after ignition
=<0.0005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text