Thermo Scientific Chemicals

Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423830250
25 L, Each
Thermo Scientific Chemicals

Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423830250
25 L, Each
Quantity
Catalog number: 423830250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-63-0
IUPAC Name
propan-2-ol
Molecular Formula
C3H8O
InChI Key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES
CC(C)O
Density
(20/20°C) 0.784 to 0.787
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
GC
>=99.5 %
Water
=<0.2 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text