Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
25 L
Catalog number 423835000
Price (USD)/ Each
54.65
Online exclusive
64.50 
Save 9.85 (15%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
54.65
Online exclusive
64.50 
Save 9.85 (15%)
Add to cart
Isopropanol, 99.6%, ACS reagent, Thermo Scientific Chemicals
Catalog number423835000
Price (USD)/ Each
54.65
Online exclusive
64.50 
Save 9.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
Molecular Weight (g/mol)60.10
View more
SpecificationsSpecification SheetSpecification Sheet
Density(20/20°C) 0.784 to 0.787
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.5 %
Water=<0.2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only