Thermo Scientific Chemicals

Lead(II) acetate trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423840250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lead(II) acetate trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423840250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423840250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6080-56-4
IUPAC Name
λ²-lead(2+) diacetate trihydrate
Molecular Formula
C4H12O7Pb
InChI Key
MCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILES
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O
Iron (Fe)
=<10 ppm
Calcium (Ca)
=<50 ppm
Potassium (K)
=<50 ppm
Insoluble matter
=<0.01 %
Nitrate and Nitrite (as NO3)
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text