Lead(II) acetate trihydrate, 99+%, ACS reagent
Lead(II) acetate trihydrate, 99+%, ACS reagent
Lead(II) acetate trihydrate, 99+%, ACS reagent
Lead(II) acetate trihydrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Lead(II) acetate trihydrate, 99+%, ACS reagent

CAS: 6080-56-4 | C4H12O7Pb | 379.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423841000
Price (USD)
37.65
Special offer
Online exclusive
43.70
Save 6.05 (14%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
37.65
Special offer
Online exclusive
43.70
Save 6.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS6080-56-4
IUPAC Nameλ²-lead(2+) diacetate trihydrate
Molecular FormulaC4H12O7Pb
InChI KeyMCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<10 ppm
Calcium (Ca)=<50 ppm
Potassium (K)=<50 ppm
Insoluble matter=<0.01 %
Nitrate and Nitrite (as NO3)=<0.005 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
How can I estimate binding capacity to Dynabeads Streptavidin magnetic beads?
What should I do if Dynabeads Streptavidin magnetic beads aggregate?
What is the definition of superparamagnetic, and what does this mean for my cell isolation application with Dynabeads magnetic beads?
What fragment length and capacity can be coupled to Dynabeads Streptavidin magnetic beads?
Can Dynabeads Streptavidin magnetic beads be boiled?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469178Certificate of AnalysisFeb 03, 2025423841000, 423840025, 423845000, 423840250
A0463278Certificate of AnalysisJan 31, 2025423841000, 423840025, 423845000, 423840250
A0468891Certificate of AnalysisJan 31, 2025423841000, 423840025, 423845000, 423840250
A0463278Certificate of AnalysisJun 26, 2024423841000, 423840025, 423845000, 423840250
A0463887Certificate of AnalysisJun 26, 2024423841000, 423840025, 423845000, 423840250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection