Thermo Scientific Chemicals

Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423850025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423850025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423850025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10099-74-8
IUPAC Name
λ²-lead(2+) dinitrate
Molecular Formula
N2O6Pb
InChI Key
RLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES
[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Form)
Crystalline powder
Titration Complexometric
>=99.0 %
Insoluble matter
=<0.005 %
Chloride (Cl)
=<10 ppm
Copper (Cu)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text