Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals
Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 423855000
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
Add to cart
Lead(II) nitrate, ACS reagent, Thermo Scientific Chemicals
Catalog number423855000
Price (USD)/ Each
92.65
Online exclusive
103.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10099-74-8
IUPAC Nameλ²-lead(2+) dinitrate
Molecular FormulaN2O6Pb
InChI KeyRLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Complexometric>=99.0 %
Insoluble matter=<0.005 %
Chloride (Cl)=<10 ppm
Copper (Cu)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only