Thermo Scientific Chemicals

Magnesium oxide, 95+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423890500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Magnesium oxide, 95+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423890500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 423890500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-48-4
IUPAC Name
oxomagnesium
Molecular Formula
MgO
InChI Key
CPLXHLVBOLITMK-UHFFFAOYSA-N
SMILES
O=[Mg]
Titration Complexometric
>=95.0 % (dried basis)
Water-soluble substances
=<0.4 %
Appearance (Color)
White
Insoluble matter
=<0.02 % (in dilute HCl)
Loss on ignition
=<2.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text