Magnesium oxide, 95+%, ACS reagent, Thermo Scientific Chemicals
Magnesium oxide, 95+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium oxide, 95+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number 423892500
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
240.00 
Save 35.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
240.00 
Save 35.35 (15%)
Add to cart
Magnesium oxide, 95+%, ACS reagent, Thermo Scientific Chemicals
Catalog number423892500
Price (USD)/ Each
204.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
240.00 
Save 35.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
Molecular Weight (g/mol)40.30
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=95.0 % (dried basis)
Water-soluble substances=<0.4 %
Insoluble matter=<0.02 % (in dilute HCl)
Loss on ignition=<2.0 %
Chloride (Cl)=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only