Magnesium sulfate heptahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Magnesium sulfate heptahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 423900250
Price (USD)/ Each
43.65
Online exclusive
48.70 
Save 5.05 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
43.65
Online exclusive
48.70 
Save 5.05 (10%)
Add to cart
Magnesium sulfate heptahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Catalog number423900250
Price (USD)/ Each
43.65
Online exclusive
48.70 
Save 5.05 (10%)
-
Add to cart
Chemical Identifiers
CAS10034-99-8
IUPAC Namemagnesium(2+) heptahydrate sulfate
Molecular FormulaH14MgO11S
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Titration Complexometric98.0 to 102.0 %
pH5.0 to 8.2 (5% soln. at 25°C)
Appearance (Color)White
Insoluble matter=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only