Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423910025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423910025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423910025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10034-96-5
IUPAC Name
manganese(2+) hydrate sulfate
Molecular Formula
H2MnO5S
InChI Key
ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES
O.[Mn++].[O-]S([O-])(=O)=O
Titration Complexometric
98.0 to 101.0 %
Heavy metals (as Pb)
=<20 ppm
Loss on ignition
10.0 % to 12.0 %
KMnO4-reducing substance
Passes test
Insoluble matter
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text