Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 423915000
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
Add to cart
Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Catalog number423915000
Price (USD)/ Each
104.65
Online exclusive
116.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10034-96-5
IUPAC Namemanganese(2+) hydrate sulfate
Molecular FormulaH2MnO5S
InChI KeyISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILESO.[Mn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric98.0 to 101.0 %
Heavy metals (as Pb)=<20 ppm
Loss on ignition10.0 % to 12.0 %
KMnO4-reducing substancePasses test
Insoluble matter=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only