Thermo Scientific Chemicals

D-Mannitol, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423920050
5 kg, Plastic drum, Each
Thermo Scientific Chemicals

D-Mannitol, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423920050
5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 423920050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69-65-8
IUPAC Name
hexane-1,2,3,4,5,6-hexol
Molecular Formula
C6H14O6
InChI Key
FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES
OCC(O)C(O)C(O)C(O)CO
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Loss on drying
=<0.05 % (5 g, 105°C, 2 hrs)
Residue after ignition
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text