Thermo Scientific Chemicals

Mercury(II) nitrate monohydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423941000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Mercury(II) nitrate monohydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423941000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 423941000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-34-8
IUPAC Name
mercury(2+) hydrate dinitrate
Molecular Formula
H2HgN2O7
InChI Key
KVICROHOONHSRH-UHFFFAOYSA-N
SMILES
O.[Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Chloride (Cl)
=<20 ppm
Sulfate (SO4)
=<20 ppm
Iron (Fe)
=<10 ppm
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text