Methanol, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Methanol, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol, 99.8+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
4 L
20 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 423950025
Price (USD)/ Each
85.65
Online exclusive
95.20 
Save 9.55 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
85.65
Online exclusive
95.20 
Save 9.55 (10%)
Add to cart
Methanol, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Catalog number423950025
Price (USD)/ Each
85.65
Online exclusive
95.20 
Save 9.55 (10%)
-
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.8 %
Water=<0.1 % (K.F.)
Density(20/4°C) 0.791 to 0.793
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only