Molybdic acid, ACS reagent, Thermo Scientific Chemicals
Molybdic acid, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdic acid, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 423985000
Price (USD)/ Each
170.65
Online exclusive
200.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
170.65
Online exclusive
200.00 
Save 29.35 (15%)
Add to cart
Molybdic acid, ACS reagent, Thermo Scientific Chemicals
Catalog number423985000
Price (USD)/ Each
170.65
Online exclusive
200.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedioxomolybdenumdiol
Molecular FormulaH2MoO4
InChI KeyVLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILESO[Mo](O)(=O)=O
Molecular Weight (g/mol)161.96
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.002 %
Phosphate (PO4)=<5 ppm
Sulfate (SO4)=<0.2 %
Heavy metals (as Pb)=<0.003 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only