Thermo Scientific Chemicals

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423995000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(1-Naphthyl)ethylenediamine, dihydrochloride, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 423995000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 423995000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1465-25-4
IUPAC Name
dihydrogen N1-(naphthalen-1-yl)ethane-1,2-diamine dichloride
Molecular Formula
C12H16Cl2N2
InChI Key
MZNYWPRCVDMOJG-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NCCNC1=C2C=CC=CC2=CC=C1
Appearance (Color)
White to beige to pink
Appearance (Form)
Powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration Argentometric
>=98.0 % (on dry substance)
Water
=<5 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text